missing translation for 'onlineSavingsMsg'
Learn More

4-(2,4-Difluorobenzoyl)piperidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128031
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39795G 5 g
D397925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39795G Supplier TCI America Supplier No. D39795G
Only null left

Chemical Identifiers

CAS 106266-04-0
Molecular Formula C12H14ClF2NO
Molecular Weight (g/mol) 261.697
MDL Number MFCD01313310
InChI Key QPJONRGTWKXJLG-UHFFFAOYSA-N
Synonym 4-2,4-difluorobenzoyl piperidine hydrochloride, 4-2,4-difluorobenzoyl piperidine hcl, 4-2,4-difluorobenzoyl-piperidine hydrochloride, 2,4-difluorophenyl piperidin-4-yl methanone hydrochloride, 2,4-difluoro-phenyl-piperidin-4-yl-methanone hydrochloride, 2,4-difluorophenyl-4-piperidinyl-methanone hcl, methanone, 2,4-difluorophenyl-4-piperidinyl-, hydrochloride, pubchem16202, acmc-1bvim, 4-2,4-difluorobenzoyl-piperidinehydrochloride
PubChem CID 18759874
IUPAC Name (2,4-difluorophenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES C1CNCCC1C(=O)C2=C(C=C(C=C2)F)F.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 261.70
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-(2,4-Difluorobenzoyl)piperidine Hydrochloride