missing translation for 'onlineSavingsMsg'
Learn More

4-(2,3-Dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid, 97%, Thermo Scientific™

Catalog No. BTB12838EA
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB12838EA 10g Amber Glass Bottle
BTB12838DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB12838EA Supplier Thermo Scientific Chemicals Supplier No. BTB12838EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54557-81-2
Molecular Formula C12H12O5
Molecular Weight (g/mol) 236.223
MDL Number MFCD00168683
InChI Key LMDXEMFSAHAGGP-UHFFFAOYSA-N
Synonym 4-2,3-dihydro-1,4-benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydro-benzo 1,4 dioxin-6-yl-4-oxo-butyric acid, 4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-4-oxobutanoic acid, 4-3,4-ethylenedioxy phenyl-4-oxobutyric acid, 4-2,3-dihydro-1,4,benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydrobenzo b 1,4 dioxin-7-yl-4-oxobutanoic acid, 4-oxo-4-2,3-dihydro-1,4-benzodioxin-6-yl butyric acid, 4-2h,3h-benzo 3,4-e 1,4-dioxin-6-yl-4-oxobutanoic acid, maybridge1_001974, regid_for_cid_2758835
PubChem CID 2758835
IUPAC Name 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid
SMILES C1COC2=C(O1)C=CC(=C2)C(=O)CCC(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10g
Formula Weight 236.22
Percent Purity 97%
Chemical Name or Material 4-(2, 3-Dihydro-1, 4-benzodioxin-6-yl)-4-oxobutanoic acid