missing translation for 'onlineSavingsMsg'
Learn More

4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid Hydrate 98.0+%, TCI America™

Catalog No. A26181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26181G 1 g
A26185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26181G Supplier TCI America Supplier No. A26181G
Only null left

Chemical Identifiers

CAS 137281-39-1
Molecular Formula C15H14N4O3
Molecular Weight (g/mol) 298.30
MDL Number MFCD11040439
InChI Key AIZPFZIKHIJCQX-UHFFFAOYSA-N
Synonym 2-Amino-4,7-dihydro-5-[2-[4-(carboxy)phenyl]ethyl]-4-oxo-3H-pyrrolo[2,3-d]pyrimidine
PubChem CID 9796279
IUPAC Name 4-(2-{2-amino-4-oxo-1H,4H,7H-pyrrolo[2,3-d]pyrimidin-5-yl}ethyl)benzoic acid
SMILES NC1=NC(=O)C2=C(NC=C2CCC2=CC=C(C=C2)C(O)=O)N1

Specifications

Quantity 1 g
Formula Weight 298.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[2-(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoic Acid Hydrate