missing translation for 'onlineSavingsMsg'
Learn More

4-([2,2':6',2″-Terpyridin]-4'-yl)benzoic Acid 98.0+%, TCI America™

Catalog No. T3405200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3405200MG 200 mg
T34051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3405200MG Supplier TCI America Supplier No. T3405200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 158014-74-5 | C22H15N3O2 | 353.381 g/mol

Chemical Identifiers

CAS 158014-74-5
Molecular Formula C22H15N3O2
Molecular Weight (g/mol) 353.381
MDL Number MFCD22056799
InChI Key RPRRPKJKACRJDD-UHFFFAOYSA-N
Synonym 4′C-(4-Carboxyphenyl)-2,2′C:6′C,2′C′C-terpyridine
PubChem CID 15903886
IUPAC Name 4-(2,6-dipyridin-2-ylpyridin-4-yl)benzoic acid
SMILES C1=CC=NC(=C1)C2=CC(=CC(=N2)C3=CC=CC=N3)C4=CC=C(C=C4)C(=O)O

Specifications

Melting Point 338°C
Color White-Yellow
Quantity 200 mg
Formula Weight 353.38
Percent Purity ≥98.0% (GC)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 4-([2,2′:6′,2′′-Terpyridin]-4′-yl)benzoic Acid