missing translation for 'onlineSavingsMsg'
Learn More

4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine 95.0+%, TCI America™

Catalog No. T35975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35975G 5 g
T35971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35975G Supplier TCI America Supplier No. T35975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 289706-46-3
Molecular Formula C9H18F3NO2Si

Specifications

Color Yellow
Quantity 5 g
Formula Weight 257.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-[2,2,2-Trifluoro-1-[(trimethylsilyl)oxy]ethyl]morpholine