missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™

Catalog No. p-3175997
Change view
Click to view available options
Quantity:
250mg
1g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC56001CB 250mg
CC56001DA 1g
CC56001EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC56001CB Supplier Thermo Scientific Chemicals Supplier No. CC56001CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 160388-53-4
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD07186451
InChI Key ZHQQRHUITAFMTC-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl benzoic acid, 4-pyrazol-1-ylmethyl-benzoic acid, 4-pyrazol-1-ylmethyl benzoic acid, benzoicacid, 4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl benzoic acid, 4-1h-pyrazol-1-yl methyl benzoic acid, 4-1h-pyrazole-1-ylmethyl benzoic acid, benzoicacid,4-1h-pyrazol-1-ylmethyl, benzoic acid, 4-1h-pyrazol-1-ylmethyl
PubChem CID 6484268
IUPAC Name 4-(pyrazol-1-ylmethyl)benzoic acid
SMILES C1=CN(N=C1)CC2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzoic acid
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.