missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)aniline, 95%, Thermo Scientific™

Catalog No. p-3522502
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC56014CB 250mg
CC56014DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC56014CB Supplier Thermo Scientific Chemicals Supplier No. CC56014CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142335-61-3
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD03422514
InChI Key ISRYTHBRUSOVAB-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl aniline, 4-1h-pyrazol-1-yl methyl aniline, 4-pyrazol-1-ylmethyl aniline, 4-1-pyrazolyl methyl aniline, 4-pyrazol-1-ylmethyl-phenylamine, 4-pyrazol-1-ylmethylphenylamine, benzenamine,4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl phenylamine, pyrazolylmethylaniline
PubChem CID 2764412
IUPAC Name 4-(pyrazol-1-ylmethyl)aniline
SMILES NC1=CC=C(CN2C=CC=N2)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250mg
Formula Weight 173.22
Percent Purity 95%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)aniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.