missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)aniline, 95%, Thermo Scientific™

Numéro de catalogue. CC56014CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC56014CB 250mg
CC56014DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC56014CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC56014CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 142335-61-3
Formule moléculaire C10H11N3
Poids moléculaire (g/mol) 173.22
Numéro MDL MFCD03422514
Clé InChI ISRYTHBRUSOVAB-UHFFFAOYSA-N
Synonyme 4-1h-pyrazol-1-ylmethyl aniline, 4-1h-pyrazol-1-yl methyl aniline, 4-pyrazol-1-ylmethyl aniline, 4-1-pyrazolyl methyl aniline, 4-pyrazol-1-ylmethyl-phenylamine, 4-pyrazol-1-ylmethylphenylamine, benzenamine,4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl phenylamine, pyrazolylmethylaniline
PubChem CID 2764412
Nom de l’IUPAC 4-(pyrazol-1-ylmethyl)aniline
SOURIRES NC1=CC=C(CN2C=CC=N2)C=C1

Spécifications

Nom ou substance chimique 4-(1H-Pyrazol-1-ylmethyl)aniline
CAS % minimum 94.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Quantité 250mg
Poids de formule 173.22
Pourcentage de pureté 95%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.