missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-yl)phenyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. p-2700467
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC18407CB 250mg Amber Glass Bottle
CC18407DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC18407CB Supplier Thermo Scientific Chemicals Supplier No. CC18407CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 352018-96-3
Molecular Formula C10H7N3S
Molecular Weight (g/mol) 201.25
MDL Number MFCD02681939
InChI Key GLAOJNMWLIFKCG-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl phenyl isothiocyanate, 1-4-isothiocyanatophenyl pyrazole, 4-1h-pyrazol-1-yl phenylisothiocyanate, 1h-pyrazole,1-4-isothiocyanatophenyl, 1h-pyrazole, 1-4-isothiocyanatophenyl, 4-pyrazolylbenzenisothiocyanate, 4-1h-pyrazole-1-yl phenyl isothiocyanate
PubChem CID 2776478
IUPAC Name 1-(4-isothiocyanatophenyl)pyrazole
SMILES S=C=NC1=CC=C(C=C1)N1C=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 201.25
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-yl)phenyl isothiocyanate
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.