missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC43316DA

Quantity 1g

 View more versions of this product

Catalog No. CC43316DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

112809-54-8
183.214
LUSFCTSUDCCYLQ-UHFFFAOYSA-N
1236449
C1=CC(=CC=C1CN2C=CN=C2)C#N
C11H9N3
MFCD08060525
4-1h-imidazol-1-ylmethyl benzonitrile, 4-imidazol-1-ylmethyl benzonitrile, 1-4-cyanobenzyl imidazole, 4-1h-imidazol-1-yl methyl benzonitrile, 1-4-cyanobenzyl-1h-imidazole, 4-imidazol-1-ylmethyl-benzonitrile, 4-1-imidazolylmethyl-benzonitrile, benzonitrile,4-1h-imidazol-1-ylmethyl, acmc-1cab3
4-(imidazol-1-ylmethyl)benzonitrile

Specifications

4-(1H-Imidazol-1-ylmethyl)benzonitrile
96.5
97%
C11H9N3
MFCD08060525
LUSFCTSUDCCYLQ-UHFFFAOYSA-N
4-(imidazol-1-ylmethyl)benzonitrile
1236449
97%
112809-54-8
100.0
Amber Glass Bottle
1g
4-1h-imidazol-1-ylmethyl benzonitrile, 4-imidazol-1-ylmethyl benzonitrile, 1-4-cyanobenzyl imidazole, 4-1h-imidazol-1-yl methyl benzonitrile, 1-4-cyanobenzyl-1h-imidazole, 4-imidazol-1-ylmethyl-benzonitrile, 4-1-imidazolylmethyl-benzonitrile, benzonitrile,4-1h-imidazol-1-ylmethyl, acmc-1cab3
C1=CC(=CC=C1CN2C=CN=C2)C#N
183.214
183.21
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents