missing translation for 'onlineSavingsMsg'
Learn More

4-(1h-Imidazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Catalog No. p-4510658
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC43304CB 250mg
CC43304DA 1g
CC43304DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC43304CB Supplier Thermo Scientific Chemicals Supplier No. CC43304CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 102432-03-1
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
InChI Key PYVCQHSLEIAQET-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-ylmethyl benzaldehyde, 4-imidazol-1-ylmethyl benzaldehyde, 4-1h-imidazol-1-yl methyl benzaldehyde, benzaldehyde,4-1h-imidazol-1-ylmethyl, acmc-20e4oe, 4-imidazolylmethyl benzaldehyde, benzaldehyde, 4-1h-imidazol-1-ylmethyl
PubChem CID 11126983
IUPAC Name 4-(imidazol-1-ylmethyl)benzaldehyde
SMILES C1=CC(=CC=C1CN2C=CN=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)benzaldehyde