missing translation for 'onlineSavingsMsg'
Learn More

4-(1h-imidazol-1-ylmethyl)aniline, Thermo Scientific™

Catalog No. CC43314CB
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC43314CB 250mg
CC43314DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC43314CB Supplier Thermo Scientific Chemicals Supplier No. CC43314CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 56643-85-7
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD06660785
InChI Key DGHAOTHIDTUSJY-UHFFFAOYSA-N
Synonym 4-imidazol-1-ylmethylphenylamine, 4-1h-imidazol-1-ylmethyl aniline, 4-imidazol-1-ylmethyl aniline, 4-1h-imidazol-1-yl methyl aniline, 4-1-imidazolyl methyl aniline, 4-imidazol-1-ylmethyl-phenylamine, 1-4-aminobenzyl imidazole, 4-imidazolylmethyl phenylamine, 1-4-aminobenzyl-1h-imidazole
PubChem CID 1265319
IUPAC Name 4-(imidazol-1-ylmethyl)aniline
SMILES NC1=CC=C(CN2C=CN=C2)C=C1

Specifications

Assay Percent Range 97%
Quantity 250mg
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)aniline