missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Benzimidazol-2-yl)aniline 98.0+%, TCI America™
$315.03 - $1099.79
Chemical Identifiers
| CAS | 2963-77-1 |
|---|---|
| Molecular Formula | C13H11N3 |
| Molecular Weight (g/mol) | 209.252 |
| MDL Number | MFCD00454395 |
| InChI Key | VQFBXSRZSUJGOF-UHFFFAOYSA-N |
| Synonym | 2-(4-Aminophenyl)benzimidazole |
| PubChem CID | 345677 |
| IUPAC Name | 4-(1H-benzimidazol-2-yl)aniline |
| SMILES | C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)N |
Chemical Identifiers
| 2963-77-1 | |
| 209.252 | |
| VQFBXSRZSUJGOF-UHFFFAOYSA-N | |
| 345677 | |
| C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)N |
| C13H11N3 | |
| MFCD00454395 | |
| 2-(4-Aminophenyl)benzimidazole | |
| 4-(1H-benzimidazol-2-yl)aniline |
Specifications
| 2963-77-1 | |
| C13H11N3 | |
| 1 g | |
| VQFBXSRZSUJGOF-UHFFFAOYSA-N | |
| 4-(1H-benzimidazol-2-yl)aniline | |
| 345677 | |
| ≥98.0% (HPLC,T) | |
| 4-(1H-Benzimidazol-2-yl)aniline |
| 250°C | |
| MFCD00454395 | |
| 2-(4-Aminophenyl)benzimidazole | |
| C1=CC=C2C(=C1)NC(=N2)C3=CC=C(C=C3)N | |
| 209.252 | |
| 209.25 | |
| Crystalline Powder |
Safety and Handling
TSCA : No