missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

$335.63 - $335.63

Chemical Identifiers

CAS 112809-25-3
Molecular Formula C10H8N4
Molecular Weight (g/mol) 184.202
MDL Number MFCD07368016
InChI Key HQLYWHSJALKYOV-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl benzonitrile, 4-1h-1,2,4-triazol-1-yl methyl benzonitrile, 4-1h-1,2,4-triazolylmethyl benzonitrile, 4-1,2,4-triazol-1-ylmethyl benzonitrile, benzonitrile, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4-triazol-1-yl methyl benzonitrile, 4-1-1,2,4-triazolyl methyl benzontrile, 4-1,2,4-triazolylmethyl benzenecarbonitrile, letrozole impurity c
PubChem CID 7537616
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzonitrile
SMILES C1=CC(=CC=C1CN2C=NC=N2)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC45716DE
SDS View Documents
Thermo Scientific Chemicals
CC45716DE
5 g
Each for $335.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

112809-25-3
184.202
HQLYWHSJALKYOV-UHFFFAOYSA-N
7537616
C1=CC(=CC=C1CN2C=NC=N2)C#N
C10H8N4
MFCD07368016
4-1h-1,2,4-triazol-1-ylmethyl benzonitrile, 4-1h-1,2,4-triazol-1-yl methyl benzonitrile, 4-1h-1,2,4-triazolylmethyl benzonitrile, 4-1,2,4-triazol-1-ylmethyl benzonitrile, benzonitrile, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4-triazol-1-yl methyl benzonitrile, 4-1-1,2,4-triazolyl methyl benzontrile, 4-1,2,4-triazolylmethyl benzenecarbonitrile, letrozole impurity c
4-(1,2,4-triazol-1-ylmethyl)benzonitrile

Specifications

112809-25-3
100.0
C10H8N4
5 g
HQLYWHSJALKYOV-UHFFFAOYSA-N
4-(1,2,4-triazol-1-ylmethyl)benzonitrile
7537616
97%
96.5
97%
MFCD07368016
4-1h-1,2,4-triazol-1-ylmethyl benzonitrile, 4-1h-1,2,4-triazol-1-yl methyl benzonitrile, 4-1h-1,2,4-triazolylmethyl benzonitrile, 4-1,2,4-triazol-1-ylmethyl benzonitrile, benzonitrile, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4-triazol-1-yl methyl benzonitrile, 4-1-1,2,4-triazolyl methyl benzontrile, 4-1,2,4-triazolylmethyl benzenecarbonitrile, letrozole impurity c
C1=CC(=CC=C1CN2C=NC=N2)C#N
184.202
184.2
4-(1H-1, 2, 4-triazol-1-ylmethyl)benzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'