missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC45701DA

 View more versions of this product

Catalog No. CC45701DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-(1H-1, 2, 4-triazol-1-ylmethyl)benzoic acid
96.5
97%
C10H9N3O2
1g
DJXUEFGFCMIONN-UHFFFAOYSA-N
4-(1,2,4-triazol-1-ylmethyl)benzoic acid
3159714
97%
160388-54-5
100.0
Amber glass bottle
MFCD07187900
4-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1h-1,2,4-triazol-1-yl methyl benzoic acid, 4-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1,2,4 triazol-1-ylmethyl-benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid hydrochloride, 4-1,2,4-triazolylmethyl benzoic acid, 4-1h-1,2,4-triazole-1-ylmethyl benzoic acid, benzoic acid,4-1h-1,2,4-triazol-1-ylmethyl, benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl
C1=CC(=CC=C1CN2C=NC=N2)C(=O)O
203.201
203.2

Chemical Identifiers

160388-54-5
203.201
DJXUEFGFCMIONN-UHFFFAOYSA-N
3159714
C1=CC(=CC=C1CN2C=NC=N2)C(=O)O
C10H9N3O2
MFCD07187900
4-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1h-1,2,4-triazol-1-yl methyl benzoic acid, 4-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1,2,4 triazol-1-ylmethyl-benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid hydrochloride, 4-1,2,4-triazolylmethyl benzoic acid, 4-1h-1,2,4-triazole-1-ylmethyl benzoic acid, benzoic acid,4-1h-1,2,4-triazol-1-ylmethyl, benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl
4-(1,2,4-triazol-1-ylmethyl)benzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep container tightly closed.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions