missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-Triazol-1-yl)benzoic acid, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC18601EA

 View more versions of this product

Catalog No. CC18601EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

4-(1H-1, 2, 4-Triazol-1-yl)benzoic acid
94.5
95%
MFCD02682060
4-1h-1,2,4-triazol-1-yl benzoic acid, 4-1,2,4 triazol-1-yl-benzoic acid, 4-1,2,4-triazol-1-yl benzoic acid, benzoic acid, 4-1h-1,2,4-triazol-1-yl, benzoic acid,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzoic acid, pubchem19582, rarechem al be 1325, buttpark 9550-80, acmc-1c5ri
C1=CC(=CC=C1C(=O)O)N2C=NC=N2
189.174
189.17
162848-16-0
100.0
C9H7N3O2
10g
FOMQQGKCPYKKHQ-UHFFFAOYSA-N
4-(1,2,4-triazol-1-yl)benzoic acid
736531
95%

Chemical Identifiers

162848-16-0
189.174
FOMQQGKCPYKKHQ-UHFFFAOYSA-N
736531
C1=CC(=CC=C1C(=O)O)N2C=NC=N2
C9H7N3O2
MFCD02682060
4-1h-1,2,4-triazol-1-yl benzoic acid, 4-1,2,4 triazol-1-yl-benzoic acid, 4-1,2,4-triazol-1-yl benzoic acid, benzoic acid, 4-1h-1,2,4-triazol-1-yl, benzoic acid,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzoic acid, pubchem19582, rarechem al be 1325, buttpark 9550-80, acmc-1c5ri
4-(1,2,4-triazol-1-yl)benzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions