missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-yl)aniline, 97%, Thermo Scientific™

$62.12 - $62.12

Chemical Identifiers

CAS 6523-49-5
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD00171470
InChI Key BLNPEJXSNSBBNM-UHFFFAOYSA-N
Synonym 4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
PubChem CID 735826
IUPAC Name 4-(1,2,4-triazol-1-yl)aniline
SMILES C1=CC(=CC=C1N)N2C=NC=N2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC18614DA
SDS View Documents
Thermo Scientific Chemicals
CC18614DA
1 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6523-49-5
160.18
BLNPEJXSNSBBNM-UHFFFAOYSA-N
735826
C1=CC(=CC=C1N)N2C=NC=N2
C8H8N4
MFCD00171470
4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
4-(1,2,4-triazol-1-yl)aniline

Specifications

6523-49-5
100.0
Amber Glass Bottle
MFCD00171470
4-1,2,4 triazol-1-yl-phenylamine, 4-1h-1,2,4-triazol-1-yl aniline, 4-1,2,4-triazol-1-yl aniline, 1-4-aminophenyl-1,2,4-triazole, triazolylaniline, 1-4'-aminophenyl-1,2,4-triazole, benzenamine, 4-1h-1,2,4-triazol-1-yl, 4-1h-1,2,4-triazol-1-yl benzenamine, 4-1,2,4-triazolyl phenylamine, pubchem3927
C1=CC(=CC=C1N)N2C=NC=N2
160.18
160.18
4-(1H-1, 2, 4-Triazol-1-yl)aniline
96.5
97%
C8H8N4
1 g
BLNPEJXSNSBBNM-UHFFFAOYSA-N
4-(1,2,4-triazol-1-yl)aniline
735826
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'