missing translation for 'onlineSavingsMsg'
Learn More

4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid, ≥90%, Thermo Scientific™

Chemical Identifiers

CAS 162046-56-2
Molecular Formula C17H23NO5
Molecular Weight (g/mol) 321.373
MDL Number MFCD06658985
InChI Key LUVXDMFUUZGFST-UHFFFAOYSA-N
Synonym 4-1-tert-butoxycarbonyl piperidin-4-yloxy benzoic acid, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy benzoic acid, 4-1-boc-4-piperidyloxy benzoic acid, 4-1-tert-butoxycarbonyl-4-piperidinyl oxy benzoic acid, 4-1-boc piperidin-4-yloxy benzoic acid, 4-4-carboxyphenoxy piperidine, n-boc protected, 4-1-t-butoxycarbonyl-4-piperidyloxy benzoic acid, 4 1-t-butoxycarbonyl 4-piperadinyl oxybenzoic acid
PubChem CID 2794649
IUPAC Name 4-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxybenzoic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OC2=CC=C(C=C2)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MO00953DA
View Documents Promotion Details
N/A 1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

162046-56-2
321.373
LUVXDMFUUZGFST-UHFFFAOYSA-N
2794649
CC(C)(C)OC(=O)N1CCC(CC1)OC2=CC=C(C=C2)C(=O)O
C17H23NO5
MFCD06658985
4-1-tert-butoxycarbonyl piperidin-4-yloxy benzoic acid, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy benzoic acid, 4-1-boc-4-piperidyloxy benzoic acid, 4-1-tert-butoxycarbonyl-4-piperidinyl oxy benzoic acid, 4-1-boc piperidin-4-yloxy benzoic acid, 4-4-carboxyphenoxy piperidine, n-boc protected, 4-1-t-butoxycarbonyl-4-piperidyloxy benzoic acid, 4 1-t-butoxycarbonyl 4-piperadinyl oxybenzoic acid
4-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxybenzoic acid

Specifications

162046-56-2
97%
MFCD06658985
LUVXDMFUUZGFST-UHFFFAOYSA-N
4-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]oxybenzoic acid
2794649
97%
97.0
C17H23NO5
4-1-tert-butoxycarbonyl piperidin-4-yloxy benzoic acid, 4-1-tert-butoxycarbonyl piperidin-4-yl oxy benzoic acid, 4-1-boc-4-piperidyloxy benzoic acid, 4-1-tert-butoxycarbonyl-4-piperidinyl oxy benzoic acid, 4-1-boc piperidin-4-yloxy benzoic acid, 4-4-carboxyphenoxy piperidine, n-boc protected, 4-1-t-butoxycarbonyl-4-piperidyloxy benzoic acid, 4 1-t-butoxycarbonyl 4-piperadinyl oxybenzoic acid
CC(C)(C)OC(=O)N1CCC(CC1)OC2=CC=C(C=C2)C(=O)O
321.373
321.37
4-{[1-(tert-Butoxycarbonyl)-4-piperidinyl]oxy}benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents