missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)piperidine 98.0+%, TCI America™

Catalog No. p-7135321
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
P19325G 5 g
P193225G 25 g
2 options

Catalog No. P19325G

Supplier: TCI America P19325G

Only null left

Chemical Identifiers

CAS 5004-07-9
Molecular Formula C9H18N2
Molecular Weight (g/mol) 154.26
MDL Number MFCD00038011
InChI Key STWODXDTKGTVCJ-UHFFFAOYSA-N
Synonym 4-1-pyrrolidinyl piperidine, 4-pyrrolidin-1-yl piperidine, 4-pyrrolidin-1-yl-piperidine, 4-pyrrolidinopiperidine, 4-1-pyrrolidinyl-piperidine, piperidine, 4-1-pyrrolidinyl, 4-pyrrolidinylpiperidine, 4-pyrrolizinopiperidine, 4-pyrrolidino-piperidine
PubChem CID 78703
IUPAC Name 4-pyrrolidin-1-ylpiperidine
SMILES C1CCN(C1)C2CCNCC2

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 5 g
Formula Weight 154.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Pyrrolidinyl)piperidine