missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzonitrile, ≥90%, Thermo Scientific™

Catalog No. CC30116DA
Change view
Click to view available options
Quantity:
1g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC30116DA 1g
CC30116EA 10g
CC30116EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC30116DA Supplier Thermo Scientific Chemicals Supplier No. CC30116DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10282-30-1
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD07368512
InChI Key ZNMSYUCZLWETII-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-yl benzonitrile, 4-1-pyrrolidinyl benzonitrile, 4-pyrrolidin-1-yl-benzonitrile, benzonitrile,4-1-pyrrolidinyl, 1-4-cyanophenyl pyrrolidine, 4-pyrrolidinobenzonitrile, 4-pyrrolidinylbenzonitrile, 4-pyrrolidinylbenzenecarbonitrile, 4-pyrrolidine-1-yl-benzonitrile
PubChem CID 4961271
IUPAC Name 4-pyrrolidin-1-ylbenzonitrile
SMILES C1CCN(C1)C2=CC=C(C=C2)C#N

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 172.23
Percent Purity 90%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.