Learn More
4-(1-Pyrrolidinyl)-1-butylamine, 98%
CAS: 24715-90-0 | C8H18N2 | 142.25 g/mol
$130.11 - $538.42
Chemical Identifiers
| CAS | 24715-90-0 |
|---|---|
| Molecular Formula | C8H18N2 |
| Molecular Weight (g/mol) | 142.25 |
| MDL Number | MFCD00192591 |
| InChI Key | LSDYCEIPEBJKPT-UHFFFAOYSA-N |
| Synonym | 4-pyrrolidin-1-yl butan-1-amine, 4-1-pyrrolidinyl-1-butanamine, 1-pyrrolidinebutanamine, 4-1-pyrrolidino butylamine, 4-pyrrolidinobutylamine, 4-1-pyrrolidinyl-1-butylamine, 1-4-aminobutyl pyrrolidine, 4-pyrrolidin-1-yl-butylamine, 4-pyrrolidinylbutylamine, 1-pyrrolidinebutylamine |
| PubChem CID | 2762762 |
| IUPAC Name | 4-pyrrolidin-1-ylbutan-1-amine |
| SMILES | NCCCCN1CCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5032303
|
Thermo Scientific Chemicals
H5032303 |
1 g |
Each for $130.11
|
|
|||||
|
AAH5032306
|
Thermo Scientific Chemicals
H5032306 |
5 g |
Each for $538.42
|
|
|||||
Description
4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines
Solubility
Soluble in water
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 24715-90-0 | |
| 142.25 | |
| LSDYCEIPEBJKPT-UHFFFAOYSA-N | |
| 2762762 | |
| NCCCCN1CCCC1 |
| C8H18N2 | |
| MFCD00192591 | |
| 4-pyrrolidin-1-yl butan-1-amine, 4-1-pyrrolidinyl-1-butanamine, 1-pyrrolidinebutanamine, 4-1-pyrrolidino butylamine, 4-pyrrolidinobutylamine, 4-1-pyrrolidinyl-1-butylamine, 1-4-aminobutyl pyrrolidine, 4-pyrrolidin-1-yl-butylamine, 4-pyrrolidinylbutylamine, 1-pyrrolidinebutylamine | |
| 4-pyrrolidin-1-ylbutan-1-amine |
Specifications
| 24715-90-0 | |
| MFCD00192591 | |
| UN2735 | |
| 4-pyrrolidin-1-yl butan-1-amine, 4-1-pyrrolidinyl-1-butanamine, 1-pyrrolidinebutanamine, 4-1-pyrrolidino butylamine, 4-pyrrolidinobutylamine, 4-1-pyrrolidinyl-1-butylamine, 1-4-aminobutyl pyrrolidine, 4-pyrrolidin-1-yl-butylamine, 4-pyrrolidinylbutylamine, 1-pyrrolidinebutylamine | |
| LSDYCEIPEBJKPT-UHFFFAOYSA-N | |
| 4-pyrrolidin-1-ylbutan-1-amine | |
| 2762762 | |
| 98% |
| C8H18N2 | |
| 1 g | |
| Air Sensitive | |
| Soluble in water | |
| NCCCCN1CCCC1 | |
| 142.25 | |
| 142.25 | |
| 4-(1-Pyrrolidinyl)-1-butylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only