missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Piperidinyl)benzeneboronic acid pinacol ester, 95%

Catalog No. AAH5192206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5192206 5 g
AAH5192203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5192206 Supplier Thermo Scientific Chemicals Supplier No. H5192206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 852227-96-4 | C17H26BNO2 | 287.21 g/mol

4-(1-Piperidinyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Piperidinyl)benzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 852227-96-4
Molecular Formula C17H26BNO2
Molecular Weight (g/mol) 287.21
MDL Number MFCD07368524
InChI Key OTOKWHGMHAAFRM-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 4-1-piperidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-piperidine, piperidine,1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl, amtb222, 4-piperidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl piperidine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl piperidine
PubChem CID 4961250
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCCC1

Specifications

Quantity 5 g
Formula Weight 287.21
Percent Purity 95%
Chemical Name or Material 4-(1-Piperidinyl)benzeneboronic acid pinacol ester
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only