Learn More
4-(1-Piperazinyl)aniline, 95%
CAS: 67455-41-8 | C10H15N3 | 177.25 g/mol
$59.14 - $568.42
Chemical Identifiers
| CAS | 67455-41-8 |
|---|---|
| Molecular Formula | C10H15N3 |
| Molecular Weight (g/mol) | 177.25 |
| MDL Number | MFCD01365906 |
| InChI Key | VAVOYRCCWLRTMS-UHFFFAOYSA-N |
| Synonym | 4-piperazin-1-yl aniline, 1-4-aminophenyl piperazine, 4-piperazinoaniline, 4-piperazin-1-yl-phenylamine, 4-1-piperazinyl aniline, benzenamine,4-1-piperazinyl, 4-piperazinylphenylamine, benzenamine, 4-1-piperazinyl, 4-piperazin-1-ylphenylamine, 1-p-aminophenyl piperazine |
| PubChem CID | 422925 |
| IUPAC Name | 4-piperazin-1-ylaniline |
| SMILES | NC1=CC=C(C=C1)N1CCNCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH55224MD
|
Thermo Scientific Chemicals
H55224MD |
250 mg |
Each for $59.14
|
|
|||||
|
AAH5522403
|
Thermo Scientific Chemicals
H5522403 |
1 g |
Each for $149.16
|
|
|||||
|
AAH5522406
|
Thermo Scientific Chemicals
H5522406 |
5 g |
Each for $568.42
|
|
|||||
Description
4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Piperazinoaniline(CAS#67455-41-8)is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Notes
Reactive with oxidizing agents. Keep at room temperature.
Chemical Identifiers
| 67455-41-8 | |
| 177.25 | |
| VAVOYRCCWLRTMS-UHFFFAOYSA-N | |
| 422925 | |
| NC1=CC=C(C=C1)N1CCNCC1 |
| C10H15N3 | |
| MFCD01365906 | |
| 4-piperazin-1-yl aniline, 1-4-aminophenyl piperazine, 4-piperazinoaniline, 4-piperazin-1-yl-phenylamine, 4-1-piperazinyl aniline, benzenamine,4-1-piperazinyl, 4-piperazinylphenylamine, benzenamine, 4-1-piperazinyl, 4-piperazin-1-ylphenylamine, 1-p-aminophenyl piperazine | |
| 4-piperazin-1-ylaniline |
Specifications
| 67455-41-8 | |
| Purple to White | |
| MFCD01365906 | |
| 4-piperazin-1-yl aniline, 1-4-aminophenyl piperazine, 4-piperazinoaniline, 4-piperazin-1-yl-phenylamine, 4-1-piperazinyl aniline, benzenamine,4-1-piperazinyl, 4-piperazinylphenylamine, benzenamine, 4-1-piperazinyl, 4-piperazin-1-ylphenylamine, 1-p-aminophenyl piperazine | |
| NC1=CC=C(C=C1)N1CCNCC1 | |
| 177.25 | |
| 177.25 | |
| Powder |
| 119°C to 123°C | |
| C10H15N3 | |
| 250 mg | |
| VAVOYRCCWLRTMS-UHFFFAOYSA-N | |
| 4-piperazin-1-ylaniline | |
| 422925 | |
| 97% | |
| 4-(1-Piperazinyl)aniline |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only