missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Methyl-4-piperidinylmethoxy)aniline, 96%

Catalog No. AAH52351MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52351MD 250 mg
AAH5235103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52351MD Supplier Thermo Scientific Chemicals Supplier No. H52351MD

CAS: 902454-26-6 | C13H20N2O | 220.316 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 902454-26-6
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.316
MDL Number MFCD09909628
InChI Key WZRKMUXDLMLRJV-UHFFFAOYSA-N
Synonym 4-1-methylpiperidin-4-yl methoxy aniline, 4-1-methyl-4-piperidinylmethoxy aniline, 4-1-methylpiperidin-4-yl methoxy phenylamine, 4-1-methyl-piperidin-4-ylmethoxy-phenylamine, 4-1-methyl-piperidin-4-ylmethoxy-phenyl amine, 4-1-methyl-4-piperidinyl methoxy benzenamine
PubChem CID 11564798
IUPAC Name 4-[(1-methylpiperidin-4-yl)methoxy]aniline
SMILES CN1CCC(CC1)COC2=CC=C(C=C2)N

Specifications

Quantity 250 mg
UN Number UN2811
Formula Weight 220.31
Percent Purity 96%
Chemical Name or Material 4-(1-Methyl-4-piperidinylmethoxy)aniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only