missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Imidazolyl)benzaldehyde 97.0+%, TCI America™

Catalog No. I08181G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08181G 1 g
I08185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08181G Supplier TCI America Supplier No. I08181G
Only null left

Chemical Identifiers

CAS 10040-98-9
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD00209977
InChI Key DCICUQFMCRPKHZ-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-yl benzaldehyde, 4-imidazol-1-yl-benzaldehyde, 4-1-imidazolyl benzaldehyde, 1-4-formylphenyl imidazole, 4-imidazol-1-yl benzaldehyde, 1-4-formylphenyl-1h-imidazole, 4-1h-imidazol-1-yl-benzaldehyde, benzaldehyde, 4-1h-imidazol-1-yl, 4-1h-imidazol-1-yl benzenecarbaldehyde, p-1-imidazolyl benzaldehyde
PubChem CID 736530
IUPAC Name 4-imidazol-1-ylbenzaldehyde
SMILES C1=CC(=CC=C1C=O)N2C=CN=C2

Specifications

Melting Point 149°C
Quantity 1 g
Formula Weight 172.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(1-Imidazolyl)benzaldehyde