missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Hydroxyethyl)pyridine, 97%

Catalog No. AAH5871106
Change view
Click to view available options
Quantity:
5 g
30 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5871106 5 g
AAH5871115 30 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5871106 Supplier Thermo Scientific Chemicals Supplier No. H5871106
Only null left

CAS: 23389-75-5 | C7H9NO | 123.155 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23389-75-5
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00077867
InChI Key HVOAMIOKNARIMR-UHFFFAOYSA-N
Synonym 1-pyridin-4-yl ethanol, 4-1-hydroxyethyl pyridine, 1-4-pyridyl ethanol, 1-pyridin-4-yl-ethanol, 1-pyridin-4-yl ethan-1-ol, 1-4-pyridinyl ethanol, +/--1-4-pyridyl ethanol, r-4-1-hydroxyethyl pyridine, alpha-methylpyridine-4-methanol, 4-pyridylethanol
PubChem CID 90919
IUPAC Name 1-pyridin-4-ylethanol
SMILES CC(C1=CC=NC=C1)O

Specifications

Melting Point 51°C to 55°C
Quantity 5 g
Formula Weight 123.15
Percent Purity 97%
Chemical Name or Material 4-(1-Hydroxyethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only