missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Cyclohexen-1-yl)morpholine, 97%

Catalog No. p-7026164
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
AAB2200414 25 g
AAB2200422 100 g
2 options

Catalog No. AAB2200414

Supplier: Thermo Scientific Chemicals B2200414

Only null left

CAS: 670-80-4 | C10H17NO | 167.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 670-80-4
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00006163
InChI Key IIQFBBQJYPGOHJ-UHFFFAOYSA-N
Synonym 4-cyclohex-1-enyl-morpholine, 1-morpholino-1-cyclohexene, 1-morpholinocyclohexene, n-morpholino-1-cyclohexene, 4-1-cyclohexen-1-yl morpholine, morpholine, 4-1-cyclohexen-1-yl, 4-cyclohex-1-en-1-yl morpholine, 4-1-cyclohexenyl morpholine, 1-morpholin-1-ylcyclohexene, 1-n-morpholino cyclohexene
PubChem CID 69589
IUPAC Name 4-(cyclohexen-1-yl)morpholine
SMILES C1CCC(=CC1)N2CCOCC2

Specifications

Density 1.02
Boiling Point 118°C to 120°C (10 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.5146
Quantity 25 g
Beilstein 118696
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 4-(1-Cyclohexen-1-yl)morpholine
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 211-579-6
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only