missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Oxazol-5-yl)benzoic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 250161-45-6
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD02682033
InChI Key MCPBXFWYUHYSIZ-UHFFFAOYSA-N
Synonym 4-1,3-oxazol-5-yl benzoic acid, 4-oxazol-5-yl benzoic acid, 4-5-oxazolyl benzoic acid, benzoic acid, 4-5-oxazolyl, 4-oxazol-5-ylbenzoic acid, 4-5-oxazolyl benzoicacid, 4-oxazol-5-yl-benzoic acid, benzoic acid,4-5-oxazolyl, 4-l,3-oxazol-5-yl benzoic acid
PubChem CID 2776414
IUPAC Name 4-(1,3-oxazol-5-yl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=CN=CO1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC14501CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

250161-45-6
189.17
MCPBXFWYUHYSIZ-UHFFFAOYSA-N
2776414
OC(=O)C1=CC=C(C=C1)C1=CN=CO1
C10H7NO3
MFCD02682033
4-1,3-oxazol-5-yl benzoic acid, 4-oxazol-5-yl benzoic acid, 4-5-oxazolyl benzoic acid, benzoic acid, 4-5-oxazolyl, 4-oxazol-5-ylbenzoic acid, 4-5-oxazolyl benzoicacid, 4-oxazol-5-yl-benzoic acid, benzoic acid,4-5-oxazolyl, 4-l,3-oxazol-5-yl benzoic acid
4-(1,3-oxazol-5-yl)benzoic acid

Specifications

250161-45-6
100.0
C10H7NO3
4-1,3-oxazol-5-yl benzoic acid, 4-oxazol-5-yl benzoic acid, 4-5-oxazolyl benzoic acid, benzoic acid, 4-5-oxazolyl, 4-oxazol-5-ylbenzoic acid, 4-5-oxazolyl benzoicacid, 4-oxazol-5-yl-benzoic acid, benzoic acid,4-5-oxazolyl, 4-l,3-oxazol-5-yl benzoic acid
OC(=O)C1=CC=C(C=C1)C1=CN=CO1
189.17
189.17
4-(1, 3-Oxazol-5-yl)benzoic acid
96.5
97%
MFCD02682033
MCPBXFWYUHYSIZ-UHFFFAOYSA-N
4-(1,3-oxazol-5-yl)benzoic acid
2776414
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents