Learn More
Sklar Instruments Scissors, Surgi-OR TC Iris Scissors, Sklar 4-1/2 in., Sharp/Sharp, Straight, Smooth, Tungsten Carbide / Stainless Steel, EA.
Catalog No. 501094039
Encompass
Scissors, Surgi-OR TC Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Tungsten Carbide / Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Hand-crafted in Pakistan, Surgi-OR TC Iris Scissors are a mid-grade instrument forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards.
Catalog No. 501094039 Supplier Sklar Instruments Supplier No. 95124
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Scissors, Surgi-OR TC Iris Scissors, Sklar, 4-1/2 in., Sharp/Sharp, Straight, Smooth, Tungsten Carbide / Stainless Steel, EA. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures. Hand-crafted in Pakistan, Surgi-OR TC Iris Scissors are a mid-grade instrument forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards.