Scissors, Surgi-OR Strabismus Scissors, Sklar, 4-1/2 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA. Surgi-OR Strabismus Scissors are an ophthalmic dissecting instrument primarily used during procedures for the treatment of Strabismus, otherwise known as in.cross-eyes. in. Hand-crafted in Pakistan, these mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. Surgi-OR instruments are a cost-effective line of instruments that must meet Sklar's quality standards. This product is curved with smooth, blunt/blunt tips and a length of 4-1/2 inches.