missing translation for 'onlineSavingsMsg'
Learn More

(3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol 98.0+%, TCI America™

Catalog No. p-7129954
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08785G 5 g
F087825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08785G Supplier TCI America Supplier No. F08785G
Only null left

Chemical Identifiers

CAS 105812-81-5
Molecular Formula C13H19FNO
Molecular Weight (g/mol) 224.30
MDL Number MFCD06658161
InChI Key CXRHUYYZISIIMT-AAEUAGOBSA-O
Synonym 3s,4r-4-4-fluorophenyl-3-hydroxymethyl-1-methylpiperidine, 3s,4r-4-4-fluorophenyl-1-methyl-3-piperidinemethanol, paroxol, unii-idr7471br7, 3s,4r-4-4-fluorophenyl-1-methylpiperidin-3-yl methanol, 3s-4-4'-fluorophenyl-3-hydroxymethyl-1-methylpiperidine, 3s,4r-4-4-fluoro-phenyl-1-methyl-piperidin-3-yl-methanol, 3-piperidinemethanol, 4-4-fluorophenyl-1-methyl-, 3s,4r, trans-4-4-fluorophenyl-3-hydroxymethyl-1-methyl-piperidine
PubChem CID 2734218
IUPAC Name (3S,4R)-4-(4-fluorophenyl)-3-(hydroxymethyl)-1-methylpiperidin-1-ium
SMILES C[NH+]1CC[C@H]([C@H](CO)C1)C1=CC=C(F)C=C1

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 223.29
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol