missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(-)-3-(Dimethylamino)pyrrolidine 98.0+%, TCI America™

Catalog No. p-7127163
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21931G 1 g
D21935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21931G Supplier TCI America Supplier No. D21931G

Chemical Identifiers

CAS 132883-44-4
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00191348
InChI Key AVAWMINJNRAQFS-LURJTMIESA-N
Synonym 3s---3-dimethylamino pyrrolidine, s-3-dimethylaminopyrrolidine, s---3-dimethylamino pyrrolidine, s-n,n-dimethylpyrrolidin-3-amine, 3s-n,n-dimethylpyrrolidin-3-amine, s-3-dimethylamino pyrrolidine, 3s-n,n-dimethyl-3-pyrrolidinamine, 3s-3-dimethylaminopyrrolidine, 3-pyrrolidinamine, n,n-dimethyl-, 3s, s-dimethyl-pyrrolidin-3-yl-amine
PubChem CID 7019156
IUPAC Name (3S)-N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCNC1

Specifications

Color Yellow
Boiling Point 164°C
Quantity 1 g
UN Number 2734
Formula Weight 114.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (3S)-(-)-3-(Dimethylamino)pyrrolidine