missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(+)-3-Aminopyrrolidine Dihydrochloride 98.0+%, TCI America™

Catalog No. A10545G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10545G 5 g
A105425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10545G Supplier TCI America Supplier No. A10545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116183-83-6
Molecular Formula C4H12Cl2N2
Molecular Weight (g/mol) 159.05
MDL Number MFCD00070605
InChI Key NJPNCMOUEXEGBL-UHFFFAOYNA-N
Synonym s-+-3-aminopyrrolidine dihydrochloride, s-pyrrolidin-3-amine dihydrochloride, 3s-+-3-aminopyrrolidine dihydrochloride, 3s-pyrrolidin-3-amine dihydrochloride, s-3-aminopyrrolidine 2hcl, s-3-aminopyrrolidine dihydrochloride, s-+-3-aminopyrrolidine 2hcl, 3-pyrrolidinamine, dihydrochloride, 3s, pubchem11156, s-+-3-aminopyrrolidinedihydrochloride
PubChem CID 13500644
IUPAC Name pyrrolidin-3-amine dihydrochloride
SMILES Cl.Cl.NC1CCNC1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 159.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3S)-(+)-3-Aminopyrrolidine Dihydrochloride