missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(-)-3-Acetamidopyrrolidine 98.0+%, TCI America™

Catalog No. A11705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11705G Supplier TCI America Supplier No. A11705G

Chemical Identifiers

CAS 114636-31-6
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00082748
InChI Key HDCCJUCOIKLZNM-UHFFFAOYNA-N
Synonym 3s---3-acetamidopyrrolidine, s-n-pyrrolidin-3-yl acetamide, n-3s-pyrrolidin-3-yl acetamide, s-3-acetamidopyrrolidine, acetamide, n-3s-3-pyrrolidinyl, n-3-pyrrolidinyl acetamide #, +-n-3r-pyrrolidin-3-yl acetamide, s-3-acetamido-pyrrolidine, 3s-3-acetamidopyrrolidine, s-n-pyrrolidin-3-ylacetamide
PubChem CID 7021471
IUPAC Name N-(pyrrolidin-3-yl)acetamide
SMILES CC(=O)NC1CCNC1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 128.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3S)-(-)-3-Acetamidopyrrolidine
TSCA No
Recommended Storage Refrigerator