missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(-)-3-Acetamidopyrrolidine, 98%

Catalog No. p-7046050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1292703 1 g
AAL1292706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1292703 Supplier Thermo Scientific Chemicals Supplier No. L1292703
Only null left

CAS: 114636-31-6 | C6H12N2O | 128.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 114636-31-6
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.18
MDL Number MFCD00082748
InChI Key HDCCJUCOIKLZNM-UHFFFAOYNA-N
Synonym 3s---3-acetamidopyrrolidine, s-n-pyrrolidin-3-yl acetamide, n-3s-pyrrolidin-3-yl acetamide, s-3-acetamidopyrrolidine, acetamide, n-3s-3-pyrrolidinyl, n-3-pyrrolidinyl acetamide #, +-n-3r-pyrrolidin-3-yl acetamide, s-3-acetamido-pyrrolidine, 3s-3-acetamidopyrrolidine, s-n-pyrrolidin-3-ylacetamide
PubChem CID 7021471
IUPAC Name N-[(3S)-pyrrolidin-3-yl]acetamide
SMILES CC(=O)NC1CCNC1

Specifications

Melting Point 52°C to 57°C
Boiling Point 70°C to 72°C (40 mmHg)
Quantity 1 g
UN Number UN3259
Beilstein 4904139
Optical Rotation −46.5° (c=1 in Ethanol)
Formula Weight 128.17
Percent Purity 98%
Chemical Name or Material (3S)-(-)-3-Acetamidopyrrolidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only