missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3S)-(-)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine, TCI America™

$126.81 - $440.02

Chemical Identifiers

CAS 147081-44-5
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD03419271
InChI Key CMIBWIAICVBURI-ZETCQYMHSA-N
Synonym s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine
PubChem CID 854071
IUPAC Name tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B29215G View Documents TCI America
B29215G
5g Each for $440.02
 
B29211G View Documents TCI America
B29211G
1g Each for $126.81
 
Specifications

Chemical Identifiers

147081-44-5
186.255
CMIBWIAICVBURI-ZETCQYMHSA-N
854071
CC(C)(C)OC(=O)N1CCC(C1)N
C9H18N2O2
MFCD03419271
s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine
tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate

Specifications

217°C
5g
C9H18N2O2
s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine
CC(C)(C)OC(=O)N1CCC(C1)N
186.255
186.26
≥98.0% (GC,T)
Yellow
147081-44-5
MFCD03419271
CMIBWIAICVBURI-ZETCQYMHSA-N
tert-butyl (3S)-3-aminopyrrolidine-1-carboxylate
854071
Clear Liquid at 20°C
(3S)-(-)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine
Safety and Handling

Safety and Handling

TSCA : No