missing translation for 'onlineSavingsMsg'
Learn More

(3S)-(-)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine 98.0+%, TCI America™

Catalog No. B29215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29215G 5 g
B29211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29215G Supplier TCI America Supplier No. B29215G

Chemical Identifiers

CAS 147081-44-5
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
MDL Number MFCD03419271
InChI Key CMIBWIAICVBURI-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopyrrolidine, s-3-amino-1-n-boc-pyrrolidine, s---1-boc-3-aminopyrrolidine, tert-butyl 3s-3-aminopyrrolidine-1-carboxylate, s-tert-butyl 3-aminopyrrolidine-1-carboxylate, s-3-amino-n-boc-pyrrolidine, s-bocap, s---1-tert-butoxycarbonyl-3-aminopyrrolidine, s-3-amino-1-boc-pyrrolidine, s---n-boc-3-aminopyrrolidine
PubChem CID 854071
IUPAC Name tert-butyl 3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(N)C1

Specifications

Color Yellow
Boiling Point 217°C
Quantity 5 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3S)-(-)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine