missing translation for 'onlineSavingsMsg'
Learn More

(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%

Catalog No. AC227950500
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227950500 50 mg Glass bottle
AC227952500 250 mg Glass bottle
AC227950010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227950500 Supplier Thermo Scientific Chemicals Supplier No. 227950500

CAS: 4272-74-6 | C14H22Cl2N2O3S | 369.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4272-74-6
Molecular Formula C14H22Cl2N2O3S
Molecular Weight (g/mol) 369.30
MDL Number MFCD00065395
InChI Key YFCUZWYIPBUQBD-ZOWNYOTGSA-N
Synonym tlck hydrochloride, tlck, s-n-7-amino-1-chloro-2-oxoheptan-3-yl-4-methylbenzenesulfonamide hydrochloride, 3s-1-chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, n-alpha-p-tosyl-l-lysine chloromethyl ketone hydrochloride, na-p-tosyl-l-lysine chloromethyl ketone hydrochloride, c14h21cln2o3s hydrochloride, tosyl-l-lysyl-chloromethane hydrochloride, l-n-5-amino-1-chloroacetyl pentyl-p-toluenesulfonamide hydrochloride, p-toluenesulfonamide, n-5-amino-1-chloroacetyl pentyl-, hydrochloride, l
PubChem CID 73093
IUPAC Name N-[(3S)-7-amino-1-chloro-2-oxoheptan-3-yl]-4-methylbenzenesulfonamide;hydrochloride
SMILES [H+].[Cl-].CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 160°C to 161°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2N(CH2)4CH(NHSO2C6H4CH3)COCH2Cl·HCl
Quantity 50 mg
Specific Rotation -9.5
Formula Weight 369.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-266-4
TSCA TSCA

RUO – Research Use Only