missing translation for 'onlineSavingsMsg'
Learn More

(3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone 98.0+%, TCI America™

Catalog No. A11245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11245G 5 g
A112425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11245G Supplier TCI America Supplier No. A11245G

Chemical Identifiers

CAS 76855-69-1
Molecular Formula C13H25NO4Si
Molecular Weight (g/mol) 287.43
MDL Number MFCD00077636,MFCD00077636
InChI Key GWHDKFODLYVMQG-UHFFFAOYNA-N
Synonym 2s,3r-3-1s-1-tert-butyldimethylsilyl oxy ethyl-4-oxoazetidin-2-yl acetate
PubChem CID 53860740
IUPAC Name 3-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-4-oxoazetidin-2-yl acetate
SMILES CC(O[Si](C)(C)C(C)(C)C)C1C(NC1=O)OC(C)=O

Specifications

Melting Point 108°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 287.43
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (3R,4R)-4-Acetoxy-3-[(R)-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone
EINECSNumber (5)-6118
TSCA No