missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(+)-3-(Trifluoroacetamido)pyrrolidine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7136740
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13691G 1 g
T136925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13691G Supplier TCI America Supplier No. T13691G
Only null left

Chemical Identifiers

CAS 141043-16-5
Molecular Formula C6H10ClF3N2O
Molecular Weight (g/mol) 218.604
MDL Number MFCD02094762
InChI Key CMZSIQCZAFAEDH-PGMHMLKASA-N
PubChem CID 42614570
IUPAC Name 2,2,2-trifluoro-N-[(3R)-pyrrolidin-3-yl]acetamide;hydrochloride
SMILES C1CNCC1NC(=O)C(F)(F)F.Cl

Specifications

Melting Point 235°C
Color White
Quantity 1 g
Formula Weight 218.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3R)-(+)-3-(Trifluoroacetamido)pyrrolidine Hydrochloride