missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(+)-3-(Dimethylamino)pyrrolidine 97.0+%, TCI America™

Catalog No. D21495G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21495G 5 g
D21491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21495G Supplier TCI America Supplier No. D21495G

Chemical Identifiers

CAS 132958-72-6
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00191347
InChI Key AVAWMINJNRAQFS-ZCFIWIBFSA-N
Synonym r-3-dimethylaminopyrrolidine, r-+-3-dimethylamino pyrrolidine, r-n,n-dimethylpyrrolidin-3-amine, 3r-+-3-dimethylamino pyrrolidine, 3r-n,n-dimethylpyrrolidin-3-amine, 3r-3-dimethylaminopyrrolidine, r-3-dimethylamino pyrrolidine, r-dmap, 3r-n,n-dimethyl-3-pyrrolidinamine, r-dimethyl-pyrrolidin-3-yl-amine
PubChem CID 2758521
IUPAC Name (3R)-N,N-dimethylpyrrolidin-3-amine
SMILES CN(C)C1CCNC1

Specifications

Color Yellow
Boiling Point 166°C
Quantity 5 g
UN Number 2734
Formula Weight 114.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (3R)-(+)-3-(Dimethylamino)pyrrolidine