missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(+)-3-Acetamidopyrrolidine 98.0+%, TCI America™

Catalog No. A11695G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A11695G Supplier TCI America Supplier No. A11695G
Only null left

Chemical Identifiers

CAS 131900-62-4
Molecular Formula C6H12N2O
Molecular Weight (g/mol) 128.175
MDL Number MFCD00082729
InChI Key HDCCJUCOIKLZNM-ZCFIWIBFSA-N
Synonym 3r-+-3-acetamidopyrrolidine, r-n-pyrrolidin-3-yl acetamide, r-3-acetamidopyrrolidine, n-3r-pyrrolidin-3-yl acetamide, acetamide, n-3r-3-pyrrolidinyl, 3r---3-acetamidopyrrolidine, 3r-3-acetamidopyrrolidine, n-3-pyrrolidinyl acetamide #, +-n-3r-pyrrolidin-3-yl acetamide, r-3-acetamido-pyrrolidine
PubChem CID 7016129
IUPAC Name N-[(3R)-pyrrolidin-3-yl]acetamide
SMILES CC(=O)NC1CCNC1

Specifications

Melting Point 58°C
Quantity 5 g
Formula Weight 128.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (3R)-(+)-3-Acetamidopyrrolidine
TSCA No
Recommended Storage Refrigerator