missing translation for 'onlineSavingsMsg'
Learn More

(3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine 98.0+%, TCI America™

Catalog No. B29201G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29201G 1 g
B29205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29201G Supplier TCI America Supplier No. B29201G

Chemical Identifiers

CAS 147081-49-0
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD03427030
InChI Key CMIBWIAICVBURI-SSDOTTSWSA-N
Synonym r-+-1-boc-3-aminopyrrolidine, r-1-boc-3-aminopyrrolidine, r-3-amino-1-n-boc-pyrrolidine, r-tert-butyl 3-aminopyrrolidine-1-carboxylate, tert-butyl 3r-3-aminopyrrolidine-1-carboxylate, r-bocap, r-+-n-boc-3-aminopyrrolidine, 3r-3-amino-1-tert-butoxycarbonyl pyrrolidine, r-n-boc-3-aminopyrrolidine, r-3-amino-n-boc-pyrrolidine
PubChem CID 854070
IUPAC Name tert-butyl (3R)-3-aminopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(C1)N

Specifications

Color Yellow
Boiling Point 244°C
Quantity 1 g
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (3R)-(+)-1-(tert-Butoxycarbonyl)-3-aminopyrrolidine