Learn More
3M™ Virtua™ Readers Eyewear
Catalog No. 19810005
Change view
Click to view available options
Magnification Power:
+2.50
Catalog No. Magnification Power
19810005 +2.50
1 options
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Catalog No. 19810005 Supplier 3M™ Supplier No. 7000127513
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
For workers who have difficulty reading small print or are engaged in fine detail work
  • Bifocal readers with three magnification diopters
  • Polycarbonate lens absorbs 99.9% UV
  • Unisex styling with lightweight design weighs less than one ounce
  • High wrap for additional coverage
  • Contoured temples for comfortable fit
  • 3M Catalog# 11513-00000-20 , 3M Catalog# 11514-00000-20 , 3M Catalog# 11515-00000-20
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Lens Coating Anti-fog
Certifications/Compliance ANSI Z87.1-2015
Lens Tint Clear
Product Type Virtual Reader Eyewear
For Use With (Application) For workers who have difficulty reading small print or are engaged in fine detail work
Magnification Power +2.50
Model 11515-00000-20
Lens Material Polycarbonate
UV Protection UV400