Learn More
3M™ Virtua™ CCS Protective Eyewear with Foam-lined Gasket
Description
3M™ Virtua™ CCS Protective Eyewear lends an athletic-inspired image in a premium line of safety glasses that offers both protection and affordability. Clear anti-fog lens features a +1.5 reading diopter to help workers who have difficulty reading small print or who are engaged in small detail work. Optional foam gasket attachment helps limit eye exposure to nuisance dust while providing additional cushioning. The large wraparound lens keeps eyes more fully protected and provides a wide viewing area.
- Contoured temples fit comfortably and snugly without pressure points but also without any looseness that causes shifting
- Impact-resistant polycarbonate lens absorbs 99.9% UV rays
- Meets the requirements of CSA Z94.3-2007 and ANSI Z87.1-2010
- Easy-to-follow instructions allow quick and easy employee evaluations within 10-15 minutes per employee
- Coverage and Gap Gauge Tools
- Educational DVD-ROM
- High-contrast stitching helps inspections for wear and damage
- Some operations may require special filter lenses, machine guards, goggles, face shields or other safety equipment
- Stain and water resistant webbing with 3M Scotchgard Protector repels liquids and resists stains
- Locking, Hardshell Carrying Case
- 3M Catalog# VC215AF , 3M Catalog# VC220AF , 3M Catalog# VC225AF
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Tint | Clear |
| Magnification Power | +1.50 |
| Model | VC215AF |