Learn More
3M™ Virtua™ CCS Protective Eyewear
Catalog No. 19164102
Change view
Click to view available options
Lens Tint:
Clear
Gray
Indoor/Outdoor Mirror
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19164102 Indoor/Outdoor Mirror
19164100 Clear
19164101 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Misuse or failure to follow warnings and instructions may result in serious potential injury, including blindness or death.
Catalog No. 19164102 Supplier 3M™ Supplier No. 7000128261
Misuse or failure to follow warnings and instructions may result in serious potential injury, including blindness or death.
Cord control system is designed to keep a corded earplug attached, untangled, and ready for use
  • Has optional foam gasket attachment
  • Foam gasket helps limit eye exposure to nuisance dust while providing additional cushioning
  • Anti-Fog polycarbonate lens absorb 99.9%UV
    • Assembly
    • Cleaning
    • Demolition
    • Electrical
    • Facility Maintenance
    • Grinding
    • Machine Operations
    • Painting
    • Remediation
    • Sanding
    • Welding
    • 3M Catalog# 11872-00000-20 , 3M Catalog# 11873-00000-20 , 3M Catalog# 11874-00000-20
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3 standards.
Specifications
Lens Coating Anti-fog
Lens Tint Indoor/Outdoor Mirror
For Use With (Application) Assembly, Cleaning, Demolition, Electrical, Facility Maintenance, Grinding, Machine Operations, Painting, Remediation, Sanding, Welding
Includes With optional foam gasket attachment
Model 11874-00000-20
Temple Style Cord Control System Temple
Lens Material Polycarbonate
Frame Material Frameless
CSA Z94.3-07, ANSI Z87.1-2010