Learn More
3M™ Versaflo™ TR-616N PAPR Assembly
Catalog No. 19268003
Change view
Click to view available options
Model TR-616N
Catalog No.
19268003 Model TR-616N
1 options
Catalog No. 19268003 Supplier 3M™ Supplier No. 7100035759
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Powered air purifying respirator provides durable protection even in harsh environments
The TR-616N assembly may be combined with 3M Versaflo branded headtops and filters or cartridges to create a complete, compact, light weight, easy-to-use PAPR for protection against certain particulates, vapors and gases. Single and four bay chargers are available to charge the battery included in the TR-616N assembly. The TR-616N assembly may be the prefered choice for working in rugged environments with airborne hazards that include particulates and gases or vapors such as fiberglass layup, pesticide manufacturing, agriculture, and demolition.
  • Assembly configured of commonly used components
  • Easy to use
  • Long run times
  • Durable belt
  • Assembly Components
    • Motor/blower (FS Cat. No. 19-268-000)
    • Length adjusting breathing tube (FS Cat. No. 19-039-502 )
    • High durability belt (FS Cat. No. 19-268-016)
    • High capacity rechargeable lithium ion battery (FS Cat. No. 19-268-022)
    • Airflow indicator (FS Cat. No. 19-268-035)
    • 3M Catalog# TR-616N
Order Info
Headtops, filters, cartridges and battery charger sold separately.
Specifications
Battery Type High Capacity Lithium Ion
Product Type PAPR Assembly, No Headtop
For Use With (Application) Pesticide manufacturing, agriculture, and demolition
Includes Motor/blower, high capacity battery, length adjusting breathing tube, high durability belt, airflow indicator
Model TR-616N