Learn More
3M™ Versaflo™ TR-614N PAPR Assembly
Catalog No. 19268002
Change view
Click to view available options
Model TR-614N
Catalog No.
19268002 Model TR-614N
1 options
Catalog No. 19268002 Supplier 3M™ Supplier No. 7100035758
Powered air purifying respirator provides durable protection even in harsh environments.
The TR-614N assembly may be combined with 3M Versaflo branded headtops and filters or cartridges to create a complete, compact, light weight, easy-to-use PAPR for protection against certain particulates, vapors and gases. Single and four bay chargers are available to charge the battery included in the TR-614N assembly. The TR-614N assembly may be the prefered choice for working in dusty environments such as pharmaceutical, dry food or dry cosmetic manufacturing.
  • Assembly configured of commonly used components
  • Easy to use and clean
  • Lightweight
  • Assembly Components
    • Motor/blower (FS Cat. No. 19-268-000)
    • Standard capacity rechargeable lithium ion battery (FS Cat. No. 19-268-020)
    • Length adjusting breathing tube (FS Cat. No. 19-039-502 )
    • Easy clean belt (FS Cat. No. 19-268-018)
    • Airflow indicator (FS Cat. No. 19-268-035)
    • 3M Catalog# TR-614N
Order Info
Headtops, filters, cartridges and battery charger sold separately.
Specifications
Battery Type Lithium-ion
Product Type PAPR Assembly, No Headtop
For Use With (Application) Pharmaceutical, dry food or dry cosmetic manufacturing
Includes Motor/blower, standard battery, length adjusting breathing tube, easy clean belt, airflow indicator
Model TR-614N