missing translation for 'onlineSavingsMsg'
Learn More

3M™ Versaflo™ TR-600 Series PAPR Belts and Extenders

Catalog No. p-4913075
Change view
Click to view available options
Product Type:
Belt
Belt Extender
For Use With (Application):
Healthcare, Pharmaceutical, Pesticide Application, Animal Processing, Laboratories, Nuclear Energy, Cosmetics, Pharmaceutical Research and Production, Recycling, Dry Foods
Healthcare, Shutdown, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
Healthcare, Shutdown, Pharmaceutical, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
19268016 Belt Healthcare, Shutdown, Pharmaceutical, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
19268017 Belt Extender Healthcare, Shutdown, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
19268018 Belt Healthcare, Pharmaceutical, Pesticide Application, Animal Processing, Laboratories, Nuclear Energy, Cosmetics, Pharmaceutical Research and Production, Recycling, Dry Foods
19268019 Belt Extender Healthcare, Pharmaceutical, Pesticide Application, Animal Processing, Laboratories, Nuclear Energy, Cosmetics, Pharmaceutical Research and Production, Recycling, Dry Foods
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19268016 Supplier 3M™ Supplier No. 7100035831
Only null left

For use with Versaflo™ TR-600 Series PAPR

TR-626 High Durability Belt

  • Comfortable belt due to ergonomic design
  • Durable construction and easy to use
  • Leather leads make it suitable for grinding
  • Part of TR-613N and TR-616N assemblies, TR-600-HIK Ready-to-Use Kit
  • Accommodates waist sizes up to 52 in.
  • Applications: Healthcare, Shutdown, Pharmaceutical, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
  • Color: Gray
  • TR-626X Belt Extender
    • Comfortable due to ergonomic design
    • Easy to use extender
    • Enables TR-626 belt to be worn comfortably, when additional belt length is needed
    • Multiple TR-626X Belt Extenders may be connected in series
    • Applications: Healthcare, Shutdown, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
    • Color: Gray
    • TR-627 Easy Clean Belt
      • Comfortable due to ergonomic design
      • Easy to use belt
      • Smooth surfaces make it easier to clean
      • Part of TR-614N and TR-617N assemblies, TR-600-ECK Ready-to-Use Kit
      • Accommodates waist sizes up to 52 in.
      • Applications: Healthcare, Pharmaceutical, Pesticide Application, Animal processing, Laboratories, Nuclear energy, Cosmetics, Pharmaceutical research and production, Recycling, Dry Foods
      • Color: Gray
      • TR-627X Belt Extender
        • Comfortable due to ergonomic design
        • Easy to use extender
        • Enables TR-627 belt to be worn comfortably, when additional belt length is needed
        • Multiple TR-627X Belt Extenders may be connected in series
        • Applications: Healthcare, Pharmaceutical, Pesticide Application, Animal processing, Laboratories, Nuclear energy, Cosmetics, Pharmaceutical research and production, Recycling, Dry Foods
        • Color: Gray
        • 3M Catalog# TR-626, TR-626X, TR-627, TR-627X

Certifications

NIOSH Approved

Specifications

Color Gray
Disposable Reusable
Product Type Belt
For Use With (Application) Healthcare, Shutdown, Pharmaceutical, Foundries, Remediation, Demolition, Aggregates, Woodworking, Casting
For Use With (Equipment) Versaflo™ TR-600 Series PAPR
Model TR-626
Quantity 1 Ea.