missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Versaflo™ TR-371+ Filter Cover
Description
TR-371+ Filter Cover holds a TR-3700 Series Filter on a TR-300+ series PAPR.
Features:
- Compatible with TR-300+ PAPR System
- Window allows users to see filter label
- Replacement part
- For particulate environments
- Sold as 1 per case
- 3M Catalog# TR-371+
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifications
Specifications
| Certifications/Compliance | NIOSH |
| Color | Beige |
| For Use With (Application) | Powder Handling |
| For Use With (Equipment) | 3M™ Versaflo™TR-300N+ Series |
| Model | TR-371+ |
| Mounting Type | Belt-mounted |
| Quantity | 1 Ea. |